4,5-Dihydroxy-3-[(2-hydroxy-5-sulfophenyl)azo]-2,7-Naphthalenedisulfonic acid trisodium salt indicator

Supplier: Ambeed
Warning

A760027-100G A760027-25G A760027-5G A760027-500G
76697-756EA 54 USD
76697-756 76697-758 76697-760 77878-602
4,5-Dihydroxy-3-[(2-hydroxy-5-sulfophenyl)azo]-2,7-Naphthalenedisulfonic acid trisodium salt indicator
4,5-Dihydroxy-3-[(2-hydroxy-5-sulfophenyl)azo]-2,7-Naphthalenedisulfonic acid trisodium salt

Precaución: for Research Use Only

Formula: C₁₆H₉N₂Na₃O₁₂S₃
MDL Number: MFCD19442652
CAS Number: 3270-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR