Ethyl all-cis-5,8,11,14,17-eicosapentaenoate 98% stabilised

Supplier: Ambeed
Warning

Synonyms: EPA Ethyl Ester, all cis-5,8,11,14,17-Eicosapentaenoic Acid Ethyl Ester, Ethyl all cis-5,8,11,14,17-Icosapentaenoate, Ethyl (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoate

Stabilization: 2000 ppm Stabilised with tocopherols

A759947-100G A759947-1G A759947-25G A759947-5G A759947-500G
76697-748EA 292 USD
76697-748 76697-750 76697-752 76697-754 77878-601
Ethyl all-cis-5,8,11,14,17-eicosapentaenoate 98% stabilised
Ethyl all-cis-5,8,11,14,17-eicosapentaenoate

Precaución: for Research Use Only

Formula: C₂₂H₃₄O₂
MW: 330.51 g/mol
MDL Number: MFCD00673476
CAS Number: 86227-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR