Boc-D-Phe-ONp 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-phenylalanine 4-nitrophenyl ester

A757365-1G A757365-25G A757365-5G
76697-742EA 10.61 USD
76697-742 76697-744 76697-746
Boc-D-Phe-ONp 98%
Boc-D-Phe-ONp

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₆
MDL Number: MFCD00069874
CAS Number: 16159-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR