Butyl hexanoate 98%

Supplier: Ambeed
Warning

Synonyms: Hexanoic acid butyl ester

A752401-100G A752401-25G A752401-500G A752401-1000G
76697-728EA 18 USD
76697-728 76697-730 76697-732 77878-593
Butyl hexanoate 98%
Butyl hexanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MDL Number: MFCD00053804
CAS Number: 626-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR