6-Fluoro-1-(methylamino)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Amifloxacin

A749227-10MG A749227-1MG A749227-25MG A749227-5MG
76697-720EA 105.03 USD
76697-720 76697-722 76697-724 76697-726
6-Fluoro-1-(methylamino)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 97%
6-Fluoro-1-(methylamino)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₉FN₄O₃
MW: 334.34 g/mol
MDL Number: MFCD00864981
CAS Number: 86393-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR