7,8-Dihydroxy-4-methylcoumarin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methyldaphnetin, 7,8-Dihydroxy-4-methyl-2H-chromen-2-one
A747962-100MG
A747962-1G
A747962-250MG
76697-708EA
25.47
USD
76697-708
76697-710
76697-712
7,8-Dihydroxy-4-methylcoumarin 98%
7,8-Dihydroxy-4-methylcoumarin
Precaución: for Research Use Only
Formula:
C₁₀H₈O₄ MW: 192.17 g/mol |
MDL Number:
MFCD00016972 CAS Number: 2107-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...