7-Ethyl-2-methyl-4-undecanol 98%

Supplier: Ambeed
Warning

Synonyms: Tetradecyl Alcohol, Tetradecanol, 7-Ethyl-2-methylundecan-4-ol

A746600-25G A746600-5G
76697-702EA 74 USD
76697-702 76697-704
7-Ethyl-2-methyl-4-undecanol 98%
7-Ethyl-2-methyl-4-undecanol

Precaución: for Research Use Only

Formula: C₁₄H₃₀O
MW: 214.39 g/mol
MDL Number: MFCD00027253
CAS Number: 103-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR