Choline iodide 98%

Supplier: Ambeed
Warning

Synonyms: Choline Iodide

A746309-10G A746309-25G A746309-100G A746309-5G
76697-692EA 15 USD
76697-692 76697-694 77880-527 77880-528
Choline iodide 98%
Choline iodide

Precaución: for Research Use Only

Formula: C₅H₁₄INO
MW: 231.08 g/mol
MDL Number: MFCD00011900
CAS Number: 17773-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR