3-Methylthiobenzamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methylbenzothioamide, Thio-m-toluamide

A738366-1G A738366-250MG A738366-25G A738366-5G
76697-646EA 11.67 USD
76697-646 76697-648 76697-650 76697-652
3-Methylthiobenzamide 98%
3-Methylthiobenzamide

Precaución: for Research Use Only

Formula: C₈H₉NS
MW: 151.23 g/mol
MDL Number: MFCD01314035
CAS Number: 2362-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR