4-(Trimethylsilyloxy)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-[(Trimethylsilyl)oxy]benzaldehyde

A738303-10G A738303-1G A738303-25G A738303-5G A738303-100G
76697-638EA 32 USD
76697-638 76697-640 76697-642 76697-644 77627-486
4-(Trimethylsilyloxy)benzaldehyde 97%
4-(Trimethylsilyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂Si
MW: 194.3025 g/mol
MDL Number: MFCD00040723
CAS Number: 1012-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR