Bis(acetonitrile)platinum dichloride 98%

Supplier: Ambeed
Warning

A737598-100MG A737598-250MG A737598-1G
76697-632EA 91 USD
76697-632 76697-634 77627-484
Bis(acetonitrile)platinum dichloride 98%
Bis(acetonitrile)platinum dichloride

Precaución: for Research Use Only

Formula: C₄H₆Cl₂N₂Pt
MDL Number: MFCD00013124
CAS Number: 13869-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR