4-Ethylphenyl acetate 96%

Supplier: Ambeed
Warning

Synonyms: Acetic Acid 4-Ethylphenyl Ester

A733421-1G A733421-25G A733421-5G
76697-608EA 14 USD
76697-608 76697-610 76697-612
4-Ethylphenyl acetate 96%
4-Ethylphenyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00026970
CAS Number: 3245-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR