β-L-Guanosine 98%

Supplier: Ambeed
Warning

A731154-50MG A731154-100MG
76697-606EA 81 USD
76697-606 77878-578
β-L-Guanosine 98%
β-L-Guanosine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O₅
MDL Number: MFCD00064103
CAS Number: 26578-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR