(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1,2,3,4-Tetrahydronaphthalen-1-ol, (S)-(+)-alpha-Tetralol

A730812-100MG A730812-1G A730812-250MG
76697-600EA 40.32 USD
76697-600 76697-602 76697-604
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol 98%
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00063006
CAS Number: 53732-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR