Ammonium iron (III) sulfate dodecahydrate 99%

Supplier: Ambeed
Warning

A730712-100G A730712-1000G A730712-25G A730712-500G
76697-598EA 7.43 USD
76697-598 77602-884 77602-886 77602-888
Ammonium iron (III) sulfate dodecahydrate 99%
Ammonium iron(III) sulphate dodecahydrate

Precaución: for Research Use Only

Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
MDL Number: MFCD00150004
CAS Number: 7783-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR