Crystal violet ≥90%

Danger

Synonyms: CI 42555

A730139-10G A730139-5G
76697-588EA 7.43 USD
76697-588 76697-590
Crystal violet ≥90%
Crystal violet

Precaución: for Research Use Only

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,04406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR