Benzyl propargyl ether 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
((Prop-2-yn-1-yloxy)methyl)benzene
A729215-1G
A729215-250MG
A729215-25G
A729215-5G
76697-580EA
6
USD
76697-580
76697-582
76697-584
76697-586
Benzyl propargyl ether 97%
Benzyl propargyl ether
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O |
MDL Number:
MFCD00483987 CAS Number: 4039-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...