Benzyl 2-(tert-butylamino)acetate 97+%

Supplier: Ambeed
Warning

A728405-100G A728405-1G A728405-25G A728405-5G
76697-572EA 689.59 USD
76697-572 76697-574 76697-576 76697-578
Benzyl 2-(tert-butylamino)acetate 97+%
Benzyl 2-(tert-butylamino)acetate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 221.29 g/mol
MDL Number: MFCD00239221
CAS Number: 343319-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR