4-tert-Butylphenyl glycidyl ether 98%

Supplier: Ambeed
Danger

A728193-10G A728193-1G A728193-5G A728193-25G
76697-566EA 35 USD
76697-566 76697-568 76697-570 77878-572
4-tert-Butylphenyl glycidyl ether 98%
4-tert-Butylphenyl glycidyl ether

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MDL Number: MFCD00005136
CAS Number: 3101-60-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR