Benzoic dithioperoxyanhydride 97%

Supplier: Ambeed
Warning

A727203-100MG A727203-1G A727203-250MG
76697-560EA 36.08 USD
76697-560 76697-562 76697-564
Benzoic dithioperoxyanhydride 97%
Benzoic dithioperoxyanhydride

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₂S₂
MW: 274.35 g/mol
MDL Number: MFCD00020686
CAS Number: 644-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR