(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone 98%

Supplier: Ambeed
Warning

Synonyms: (S)-4-Isopropyl-3-propionyloxazolidin-2-one

A726497-1G A726497-250MG A726497-5G
76697-554EA 53.05 USD
76697-554 76697-556 76697-558
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone 98%
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD00043401
CAS Number: 77877-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR