Bis(phenylsulfonyl)sulfide 97%

Supplier: Ambeed
Warning

Synonyms: Benzenesulphonic thioanhydride

A725768-1G A725768-250MG A725768-5G
76697-552EA 74 USD
76697-552 77878-568 77878-569
Bis(phenylsulfonyl)sulfide 97%
Bis(phenylsulfonyl)sulfide

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₄S₃
MW: 314.388 g/mol
MDL Number: MFCD00094733
CAS Number: 4388-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR