cis-1,4-Cyclohexanediamine 95%

Supplier: Ambeed
Danger

Synonyms: cis-1,4-Diaminocyclohexane, cis-Cyclohexane-1,4-diamine

A724427-100MG A724427-1G A724427-25G A724427-5G
76697-536EA 25.47 USD
76697-536 76697-538 76697-540 76697-542
cis-1,4-Cyclohexanediamine 95%
cis-1,4-Cyclohexanediamine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD08276323
CAS Number: 15827-56-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR