Boc-D-Glu-OMe 98%

Supplier: Ambeed
Warning

A723762-1G A723762-250MG A723762-25G
76697-532EA 25 USD
76697-532 76697-534 77878-566
Boc-D-Glu-OMe 98%
Boc-D-Glu-OMe

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₆
MDL Number: MFCD00076930
CAS Number: 55227-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR