ɑ-Cyano-4-hydroxycinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-3-(4-hydroxyphenyl)acrylic Acid, α-Cyano-4-hydroxycinnamic Acid, α-CHCA, CHCA

A722869-10G A722869-1G A722869-250MG A722869-25G A722869-50MG A722869-5G
76697-514EA 66.84 USD
76697-514 76697-516 76697-518 76697-520 76697-522 76697-524
ɑ-Cyano-4-hydroxycinnamic acid 98%
alpha-Cyano-4-hydroxycinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00004204
CAS Number: 28166-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR