2',5'-Dideoxyadenosine 98+%

Supplier: Ambeed
Warning

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-methyltetrahydrofuran-3-ol

A722266-10MG A722266-1MG A722266-50MG A722266-5MG
76697-506EA 111.4 USD
76697-506 76697-508 76697-510 76697-512
2',5'-Dideoxyadenosine 98+%
2',5'-Dideoxyadenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O₂
MW: 235.2425 g/mol
MDL Number: MFCD00210904
CAS Number: 6698-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR