Butyl isovalerate 98%

Supplier: Ambeed
Warning

Synonyms: Isovaleric acid butyl ester, Butyl 3-methylbutanoate

A721266-100G A721266-25G A721266-500G
76697-488EA 14 USD
76697-488 76697-490 76697-492
Butyl isovalerate 98%
Butyl isovalerate

Precaución: for Research Use Only

Formula: C₉H₁₈O₂
MDL Number: MFCD00048775
CAS Number: 109-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR