6-(tert-Butyl)-4-methyl-2H-chromen-2-one 95%

Supplier: Ambeed
Warning

A719991-100MG A719991-1G A719991-250MG A719991-5G
76697-480EA 17 USD
76697-480 76697-482 76697-484 76697-486
6-(tert-Butyl)-4-methyl-2H-chromen-2-one 95%
6-(tert-Butyl)-4-methyl-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₄H₁₆O₂
MDL Number: MFCD00016968
CAS Number: 17874-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR