Diallylamine hydrochloride 45-55% in water

Supplier: Ambeed
Warning

A717548-100G A717548-1G A717548-25G A717548-500G A717548-5G
76697-456EA 272 USD
76697-456 76697-458 76697-460 76697-462 76697-464
Diallylamine hydrochloride 45-55% in water
Diallylamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁N
MW: 133.62 g/mol
MDL Number: MFCD00060150
CAS Number: 6147-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR