tetra-Chloroauric(III) acid hydrate 99%

Supplier: Ambeed
Danger

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

A716090-1G A716090-100MG A716090-250MG A716090-5G
76697-436EA 83 USD
76697-436 77602-828 77602-830 77602-832
tetra-Chloroauric(III) acid hydrate 99%
tetra-Chloroauric(III) acid hydrate

Precaución: for Research Use Only

Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
MDL Number: MFCD00149903
CAS Number: 27988-77-8
UN: 3260
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR