Bromo(1,10-phenanthroline)(triphenylphosphine)copper(I) 98%

Supplier: Ambeed
Warning

A715737-1G A715737-250MG A715737-5G
76697-432EA 49 USD
76697-432 76697-434 77627-464
Bromo(1,10-phenanthroline)(triphenylphosphine)copper(I) 98%
Bromo(1,10-phenanthroline)(triphenylphosphine)copper(I)

Precaución: for Research Use Only

Formula: C₃₀H₂₄BrCuN₂P
MW: 586.9487 g/mol
MDL Number: MFCD07369032
CAS Number: 25753-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR