1-Iodo-2,3,5,6-tetramethylbenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3,5,6-Tetramethyliodobenzene, 3-Iodo-1,2,4,5-tetramethylbenzene
A715135-100G
A715135-1G
A715135-25G
A715135-5G
76697-422EA
460.44
USD
76697-422
76697-424
76697-426
76697-428
1-Iodo-2,3,5,6-tetramethylbenzene 98%
1-Iodo-2,3,5,6-tetramethylbenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₃I MW: 260.12 g/mol |
MDL Number:
MFCD00013705 CAS Number: 2100-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...