Boc-β-t-butyl-L-alanine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-β-tBu-Ala-OH, (S)-2-((tert-Butoxycarbonyl)amino)-4,4-dimethylpentanoic acid, Boc-β-tBuAla-OH

A714979-100MG A714979-1G A714979-250MG A714979-5G
76697-414EA 48.81 USD
76697-414 76697-416 76697-418 76697-420
Boc-β-t-butyl-L-alanine 95%
Boc-beta-t-butyl-L-alanine

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
MDL Number: MFCD00237540
CAS Number: 79777-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR