2-(Phenyl(phenylamino)methyl)cyclohexanone 95%

Supplier: Ambeed
Warning

A711879-1G A711879-250MG
76697-406EA 99 USD
76697-406 76697-408
2-(Phenyl(phenylamino)methyl)cyclohexanone 95%
2-(Phenyl(phenylamino)methyl)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO
MW: 279.37 g/mol
MDL Number: MFCD00488518
CAS Number: 737-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR