4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde 95+%

Supplier: Ambeed
Warning

A710658-1G A710658-5G
76697-402EA 24.41 USD
76697-402 76697-404
4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde 95+%
4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₃
MW: 190.00 g/mol
MDL Number: MFCD04039026
CAS Number: 160068-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR