7-Methylimidazo[1,2-a]pyridine 96%

Supplier: Ambeed
Warning

A709581-1G A709581-250MG A709581-5G
76697-384EA 71.09 USD
76697-384 76697-386 76697-388
7-Methylimidazo[1,2-a]pyridine 96%
7-Methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
MDL Number: MFCD06761695
CAS Number: 874-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR