Carboxymethoxylamine hemihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Aminooxyacetic acid hemihydrochloride, O-(Carboxymethyl)hydroxylamine Hemihydrochloride, O-(Carboxymethyl)hydroxylamine hemihydrochloride, (Aminooxy)acetic Acid Hemihydrochloride

A709258-1G A709258-25G A709258-5G A709258-100G
76697-370EA 6 USD
76697-370 76697-372 76697-374 77627-452
Carboxymethoxylamine hemihydrochloride 95%
Carboxymethoxylamine hemihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂O₆
MDL Number: MFCD00012955
CAS Number: 2921-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR