Methyl 3,6-dichloropyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dichloro-2-(methoxycarbonyl)pyridine, Methyl 3,6-dichloropicolinate

A708751-250MG A708751-5G A708751-1G A708751-25G
76697-366EA 15 USD
76697-366 76697-368 77627-450 77880-466
Methyl 3,6-dichloropyridine-2-carboxylate 97%
Methyl 3,6-dichloropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD00186642
CAS Number: 1532-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR