Methyl hydrogen azelate 97%

Supplier: Ambeed
Warning

Synonyms: Nonanedioic acid monomethyl ester, Monomethyl Nonanedioate, Methyl Hydrogen Azelate, Monomethyl Azelate, mono-Methyl azelate, Azelaic acid monomethyl ester

A707942-1G A707942-5G A707942-25G A707942-100G
76697-362EA 7 USD
76697-362 76697-364 77627-448 77878-548
Methyl hydrogen azelate 97%
Methyl hydrogen azelate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
MDL Number: MFCD00004431
CAS Number: 2104-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR