(E)-Ethyl 2-cyano-3-phenylacrylate 95%

Supplier: Ambeed
Warning

A704285-1G A704285-25G A704285-5G
76697-336EA 19 USD
76697-336 76697-338 76697-340
(E)-Ethyl 2-cyano-3-phenylacrylate 95%
(E)-Ethyl 2-cyano-3-phenylacrylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MDL Number: MFCD00009137
CAS Number: 2169-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR