Cinnamyl bromide 98%

Supplier: Ambeed
Danger

A703815-25G A703815-5G
76697-332EA 53 USD
76697-332 76697-334
Cinnamyl bromide 98%
Cinnamyl bromide

Precaución: for Research Use Only

Formula: C₉H₉Br
MDL Number: MFCD00000245
CAS Number: 4392-24-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR