2-Amino-5-methylbenzothiazole 95%

Supplier: Ambeed
Warning

A703511-100MG A703511-250MG A703511-1G
76697-328EA 65.78 USD
76697-328 76697-330 77308-164
2-Amino-5-methylbenzothiazole 95%
2-Amino-5-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₈N₂S
MW: 164.23 g/mol
MDL Number: MFCD00205354
CAS Number: 14779-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR