3-Pyridin-2-ylacrylic acid 98%

Supplier: Ambeed
Warning

A702486-10G A702486-1G A702486-25G A702486-5G
76697-320EA 67 USD
76697-320 76697-322 76697-324 76697-326
3-Pyridin-2-ylacrylic acid 98%
3-Pyridin-2-ylacrylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MDL Number: MFCD00179422
CAS Number: 7340-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR