1-Carbamoylcyclobutane-1-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Cyclobutane-1,1-dicarboxylic acid monoamide

A700317-1G A700317-250MG
76697-314EA 41.38 USD
76697-314 76697-316
1-Carbamoylcyclobutane-1-carboxylic acid 97%
1-Carbamoylcyclobutane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.141 g/mol
MDL Number: MFCD04114274
CAS Number: 845621-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR