2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone 96%

Supplier: Ambeed
Danger

Synonyms: 2,2,2-Trifluoro-1-(4-(trifluoromethyl)phenyl)ethanone, 4-(Trifluoroacetyl)benzotrifluoride, 2,2,2-Trifluoro-1-[4-(trifluoromethyl)phenyl]ethan-1-one

A699974-1G A699974-5G
76697-310EA 30.77 USD
76697-310 77627-428
2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone 96%
2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₄F₆O
MW: 242.118 g/mol
MDL Number: MFCD00061276
CAS Number: 74853-66-0
UN: 1224
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR