N-Carbobenzoxy-1,4-diaminobutane hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: N-Z-1,4-Butanediamine hydrochloride, N-Cbz-1,4-diaminobutane Hydrochloride, Benzyl N-(4-Aminobutyl)carbamate Hydrochloride, Benzyl (4-aminobutyl)carbamate hydrochloride, N-(4-Aminobutyl)carbamic Acid Benzyl Ester Hydrochloride

A699519-1G A699519-250MG A699519-5G A699519-25G
76697-306EA 28 USD
76697-306 76697-308 77602-774 77627-426
N-Carbobenzoxy-1,4-diaminobutane hydrochloride 97%
N-Carbobenzoxy-1,4-diaminobutane hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 258.75 g/mol
MDL Number: MFCD00270149
CAS Number: 18807-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR