Biapenem 98%

Supplier: Ambeed
Warning

Synonyms: (4R,5S,6S)-3-((6,7-Dihydro-5H-pyrazolo[1,2-a][1,2,4]triazol-4-ium-6-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

A699164-100MG A699164-10MG A699164-1MG A699164-50MG A699164-5G A699164-1G A699164-250MG A699164-25G
76697-292EA 179.3 USD
76697-292 76697-294 76697-296 76697-298 77602-772 77627-420 77627-422 77627-424
Biapenem 98%
Biapenem

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₄O₄S
MW: 350.39 g/mol
MDL Number: MFCD00864863
CAS Number: 120410-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR