Copper(I) thiocyanate 98%

Supplier: Ambeed
Danger

Synonyms: Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide

A698548-100G A698548-25G A698548-500G
76697-284EA 15 USD
76697-284 76697-286 76697-288
Copper(I) thiocyanate 98%
Copper(I) thiocyanate

Precaución: for Research Use Only

Formula: CCuNS
MW: 121.63 g/mol
MDL Number: MFCD00010980
CAS Number: 1111-67-7
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,02699

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR