Methyl docosanoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl behenate, Docosanoic acid methyl ester, Behenic acid methyl ester

A694195-25G A694195-500G A694195-100G
76697-264EA 12 USD
76697-264 77573-248 77602-762
Methyl docosanoate 98%
Methyl docosanoate

Precaución: for Research Use Only

Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
MDL Number: MFCD00009347
CAS Number: 929-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR