Boc-O-benzyl-L-β-homothreonine 95+%

Supplier: Ambeed
Warning

Synonyms: (3R,4R)-4-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid

A691115-100MG A691115-1G A691115-250MG
76697-258EA 19.1 USD
76697-258 76697-260 76697-262
Boc-O-benzyl-L-β-homothreonine 95+%
Boc-O-benzyl-L-beta-homothreonine

Precaución: for Research Use Only

Formula: C₁₇H₂₅NO₅
MW: 323.39 g/mol
MDL Number: MFCD01862935
CAS Number: 254101-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR